Anti-DICER1 Rabbit Polyclonal Antibody

Supplier: Abgent
AP1901B
60941-370EA 763.01 USD
60941-370
Anti-DICER1 Rabbit Polyclonal Antibody
Antibodies
Primary Rabbit Anti-Dicer1 (C-term) Reacts with Human

Type: Primary
Antigen: DICER1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR