Anti-KAT5 Rabbit Polyclonal Antibody

Supplier: Abgent
AP1081A
60941-200EA 801.6 USD
60941-200
Anti-KAT5 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: KAT5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR