VWR® PCR Films, PE
Supplier: VWR International
60941-122
60941-122CS 109.51 USD
60941-122
VWR® PCR Films, PE
Seals Microplate Seals
These medium-density polyethylene films, 76µm thick with a 46µm thick adhesive layer, are designed for general-purpose microplate sealing and PCR.
  • Pierceable
  • Nonsterile
Films are pierceable and supplied nonsterile.