VWR® Plate Roller
Supplier: VWR International
RLPLTVWR01
60941-118CS 63.66 USD
60941-118
VWR® Plate Roller
Film Applicators Sealing Film Applicators
This 10.2 cm (4") soft rubber roller is designed for assuring tight and consistent application of adhesive films and foils to microplates.
  • Tight and consistent application
  • Secure seal around all microplate wells