VWR® Clear-Zone Pierceable Films for Robotics
Supplier: VWR International
60941-090
60941-090CS 168.41 USD
60941-090
VWR® Clear-Zone Pierceable Films for Robotics
Seals Microplate Seals
With a polyethylene top layer and inert white polypropylene and acrylic adhesive sublayer, this easy-piercing sealing film allows direct sample recovery with single or multichannel pipettors and robotic probes.
  • Clear zone above each well is free of adhesive
  • Minimise sample exposure to adhesive
  • Dimensions: 14.3x7.9 cm (55/8x31/8")
  • Recommended temperature range −40 to +90 °C
The adhesive-free zone above each well prevents adhesive fouling of the pipette tip or robotic probe, minimising the amount of adhesive that comes into contact with the sample. Suitable for 96-well plates only.