SP Bel-Art Electrophoresis Gel Plate Drying Rack, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
F13595-0000
60917-250EA 179.97 USD
60917-250
SP Bel-Art Electrophoresis Gel Plate Drying Rack, Bel-Art Products, a part of SP
Racks Electrophoresis Plate Racks
This sturdy, A-frame shaped rack holds glass electrophoresis plates upright for quick drying and convenient access.
  • Steel construction with epoxy coating
  • Space-saving design
  • Nonskid rubber pads
Rack has ten positions including three slots for large and double plates
Rubber pads on the rack base prevent sliding. Made of steel construction with baked-on epoxy coating, this space saver holds plates with a minimum width of 16.5 cm (6¹/₂”).
All slots have soft plastic supports on each side to cushion plates.