SP Bel-Art Sterileware® Scoop An' Bag™ Sampler, Sterile Polystyrene, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
H36910-0000
60917-207CS 179.1 USD
60917-207
SP Bel-Art Sterileware® Scoop An' Bag™ Sampler, Sterile Polystyrene, Bel-Art Products, a part of SP
Scoops
Sample collection system includes a sterile 60 mL (2oz.) sampling scoop and sterile 11.4x22.9 cm (4½x9”), 540 mL (18.3oz.) volume polyethylene twirl-type bag with large label area to hold the sample.
  • Sterile polystyrene scoop construction
  • Provided in polyethylene bag
  • Lot stamped for accurate lot tracing
  • Scoop features a flat bottom
The one-piece scoop is made from white, high-impact, FDA-grade polystyrene plastic, with a long handle to keep the hand away from the sample.
A flat bottom allows the scoop to remain upright for use as a weighing boat or to be placed on a flat surface. Included are 50 individually sealed and sterilized wrapped sets per box, all ready for one-time use and disposal.
Lot stamped for accurate lot tracing and shipped with a Certificate of Processing for Sterilization.