SP Bel-Art 12-Channel Mini-Tube Strips, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
F37857-0000
60917-202CS 550.97 USD
60917-202
SP Bel-Art 12-Channel Mini-Tube Strips, Bel-Art Products, a part of SP
Tubes Microtubes
These 1 mL volume Mini-Tubes offer a convenient and economical means of performing laboratory procedures using multi-channel pipettors of four, eight, or twelve channels.
  • Convenient vessel for laboratory procedures
  • Economical
  • Easily separated into single tubes
They feature an attachment bridge between tubes, forming a Mini-Tube strip which can be easily separated into single tubes by cutting apart.