VWR® Stackable Petri Dish Incubation Tray
Supplier: VWR International
60914-210
60914-210PK 98.49 USD
60914-210
VWR® Stackable Petri Dish Incubation Tray
Racks Incubator Racks
Stackable Petri dish Incubation Trays increase overall capacity of an incubator by holding five 100mm diameter or fifteen 60mm diameter Petri dishes.
  • Holes in tray bottom facilitate air flow
  • Polypropylene construction can be disinfected with 70% Ethanol or common laboratory disinfectants
  • Not recommended for use above 100°C (212°F)
  • Autoclaving may warp
Each tray includes two large label areas for critical sample identification by marker or label.