Contact Plates, Simport®
Supplier: Simport Scientific
D210-17
60872-312CS 487.69 USD
60872-312
Contact Plates, Simport®
Culture Dishes Petri Dishes
Contact plates are free from optical distortion and are supplied sterile.
  • Polystyrene construction
  • 10 x 10 alphanumeric grid facilitates counting and identification