Square Petri Dish with Grid, Simport
Supplier: Simport Scientific
D210-16NS D210-16
60872-480CS 470.73 USD
60872-480 60872-310
Square Petri Dish with Grid, Simport
Culture Dishes Petri Dishes
These square petri dishes are free from optical distortion.
They are ideal for phage typing, susceptibility testing, plate counts, and probe assats. Dishes feature an alphanumeric grid with four venting ribs on the underside of the lid to prevent build-up of condensation. Dishes are available sterile or nonsterile.