B352 Specimen Bottles and Caps, Simport Scientific
Supplier: Simport Scientific
B352
60872-302CS 1267.9 USD
60872-302
B352 Specimen Bottles and Caps, Simport Scientific
Bottles Drosophila Bottles
Polypropylene bottle useful as a Drosophila stock bottle.
  • Autoclavable
  • Graduated from 0 to 170 ml and from 0 to 6 oz.
Bottles feature a dip-stick well for small volume testing and a pour spout for dripless pouring. The square base makes this bottle very stable.
Ordering information: Snap cap must be purchased separately.