KIMAX® Culture Tubes, Reusable, Graduated, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
45071-10
60869-001CS 568.75 USD
60869-001
KIMAX® Culture Tubes, Reusable, Graduated, Kimble Chase
Tubes Culture Tubes
KIMAX® culture tube intended primarily for use in the testing of milk for the presence of bacteria by the methylene blue reductase test.
  • Smooth, fire-polished rim
  • Graduation line is located at the 10 ml level
  • Durable white ceramic enamel marking spot
Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type 1 and ASTM E438, Type 1, Class A requirements.