PYREX® Disposable Culture Tubes, Rimless, Dispenser Pack and Bulk Pack, Corning
Supplier: Corning
99445-10 99445-12 99445-13 99445-15 99445-18 99445-20 99445-16 99445-16X 99445-16XX
60826-825CS 142.15 USD
60826-825 60826-836 60826-847 60826-858 60826-891 60826-905 60826-869 60826-870 60826-880
PYREX® Disposable Culture Tubes, Rimless, Dispenser Pack and Bulk Pack, Corning
Tubes Culture Tubes
Made of borosilicate glass to reduce pH changes and contaminants potentially leached from soda-lime glass.
These one-use culture tubes are packaged for your convenience in easy-to-open, easy-to-use packs. The three small sizes come in a dispenser arrangement, and the larger sizes are bulk-packed.