Westergren Erythrocyte Sedimentation Rate Tubes, KIMBLE CHASE®, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
67G830SO
60826-605CS 627.41 USD
60826-605
Westergren Erythrocyte Sedimentation Rate Tubes, KIMBLE CHASE®, DWK Life Sciences
Clinical Diagnostic Supplies Clinical Sample Collection/Preparation Blood Sample Collection/Preparation
ESR tubes measure the rate at which red blood cells settle out of plasma.
  • Calibrated in yellow
  • Cotton plugged
Westergren sedimentation rate tubes may be used for standard procedures, by drawing blood in a liquid sodium citrate vacuum tube, or the modified procedure, using a saline-diluted EDTA vacuum tube.
Precision-drawn glass and accurate calibrations ensure that the results are accurate with standard normal values.
Certifications: Manufactured from 90 expansion soda lime glass conforming to USP Type III requirements.