Kimble® Culture Tubes, Disposable, Borosilicate Glass, DWK Life Sciences
Supplier: Kimble Chase
60T1685B 73500-1075 73500-1275 73500-16125 73500-16100 73500-20150 73500-16150 73500-18150 73500-650 73500-13100 73500-25150
33501-952CS 143.49 USD
33501-952 60825-500 60825-502 60825-504 60825-510 60825-730 60825-979 60825-980 60819-854 60825-503 89003-438
Kimble® Culture Tubes, Disposable, Borosilicate Glass, DWK Life Sciences
Tubes Culture Tubes
Culture tubes are constructed of KIMAX-51® borosilicate glass for excellent chemical durability and resistance against sodium leaching.
Tubes feature sturdy, uniform bottoms and consistent lengths. The 6×50 mm tube is also known as a 'Durham' tube. The 10×75 and 12×75 mm tubes are suitable for cell washing procedures. Tubes are packed in convenient shrink-wrapped trays for ease of use on bench tops and in drawers, as well as to eliminate breakage and contamination.
Certifications: Glass meets ASTM E438, Type I, Class B requirements. Designed from ASTM E890 specifications.