Accessories for Capillary Tubes, Globe Scientific
Supplier: Globe Scientific
51601 51674
60820-114PK 98.26 USD
60820-114 10022-158 10022-159
Accessories for Capillary Tubes, Globe Scientific
Clinical Diagnostic Supplies
Pipette filler bulbs are designed for use on all Globe Scientific capillary tubes.
Capillary tube sealant contains a unique wax compound that quickly forms a leak-tight seal. Non-toxic, non-drying, and non-hardening, the sealant is formulated to withstand centrifugation. There is no need for plastic caps or heat sealing.
They help expel the blood from the capillary tube (if needed). They are also used when drawing blood directly from a 13mm or 16mm blood collectioon tube (instead of directly from a finger).