Polypropylene Test Tube, 13×75, Globe Scientific
Supplier: Globe Scientific
110471
60819-968PK 87.11 USD
60819-968
Polypropylene Test Tube, 13×75, Globe Scientific
Tubes Test Tubes
Polypropylene tubes can withstand 3000 rpm.
They are autoclavable and freezable.