Semi-quantitative test strips QUANTOFIX Nitrate / Nitrite
Supplier: Macherey-Nagel
91313
60787-738PK 89.28 USD
60787-738
Semi-quantitative test strips QUANTOFIX Nitrate / Nitrite
Environmental Test Kits
For semi-quantitative determinations
Gradation: 0 to 500mg/L. Color changes from white to red/violet.