Semi-quantitative test strips QUANTOFIX Sulfite
Supplier: Macherey-Nagel
91306
60787-728PK 183.62 USD
60787-728
Semi-quantitative test strips QUANTOFIX Sulfite
Environmental Test Kits
For semi-quantitative determinations
Gradation: 0 to 1000mg/L. Color changes from white to salmon.