Semi-quantitative test strips QUANTOFIX Iron
Supplier: Macherey-Nagel
91330
60787-724PK 45.87 USD
60787-724
Semi-quantitative test strips QUANTOFIX Iron
Environmental Test Kits
For semi-quantitative determinations
Gradation: 0 to 1000mg/L. Color changes from white to dark