Hydrion™, Mikro pH Test Paper with Dispensor, Micro Essential Laboratory
Supplier: Micro Essential
M-1200
60775-881CS 147.99 USD
60775-881
Hydrion™, Mikro pH Test Paper with Dispensor, Micro Essential Laboratory
Test Paper pH Indicator Papers and Test Strips
Hydrion® Double Roll pH Paper set the standard for quality, accuracy, and reliability
  • Composed of Special Wet Strength Paper
  • Color chart provides clear color matches based on spectral color changes for sample comparison
  • These Mikro paper rolls measure 180 x 5/32 inches
These rolls test the 1.0-12.0 pH range with each full pH unit showing a distinct color variation.
Packaging: Packaged in a dual roll protective plastic dispenser and contained in a polypropylene case to guard against sunlight and laboratory hazards.