SP Bel-Art FLUO-KEM® Teflon® Resin Lab-Thread Tape, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
F24020-0000
60490-100CS 261.72 USD
60490-100
SP Bel-Art FLUO-KEM® Teflon® Resin Lab-Thread Tape, Bel-Art Products, a part of SP
Tapes
Pure, non-sintered Teflon® PTFE Lab-Thread Tape contains no adhesive, yet does a superior job of sealing small apertures.
  • Non-sintered
  • Seals small apertures securely without adhesive
  • Withstands a wide range of temperatures
  • No need for lubricants and vacuum greases
Conforming to irregularities of surfaces, threads and joints of all types, it eliminates the need for lubricants and vacuum greases
Can be used on ground glass joints, desiccators, bell jars, freezedrying equipment, etc. 0.0635 mm (0.0025") thick, 12.7 mm (½") wide x 7.6 meters long (300") per roll. Withstands temperatures from -250ºF to 500ºF.
Provides a vacuum tight seal and eliminates ˝freezing˝ even after long periods.