Disposable Tip Adapter, Hamilton Company
Supplier: Hamilton
31331
60384-079EA 324.31 USD
60384-079
Disposable Tip Adapter, Hamilton Company
Syringes Syringes for Chromatography
Hamilton company offers the most comprehensive selection of standard and custom syringes and accessories in the industry
Hamilton disposable tip adapters enable the connection of a standard 1 mL or 200 uL disposable pipette tip to the end of a Hamilton syringe. The adapter connects to standard Luer Tip (LT) and PTFE Luer Lock (TLL) syringes. When used together, the PB-600 and the disposable tip adapters can be used as a small volume repeating pipette. The adapter is ideal for eliminating the possibility for cross contamination.
Each is manufactured to achieve the highest level ofMore Product Information