SN Barrel/Plunger Assembly without Needle, for Model 802
Supplier: HAMILTON COMPANY
Model: 802. Volume: 25 uL. Reinforced plungers. Cemented Needles (N) or removable Needles (RN). Plunger barrel assemblies are replaceable. Plunger stop prevents plunger blow-out.