HT8 GC Capillary Columns, SGE
Supplier: Trajan Scientific and Medical
054677 054682 054680 054676 054675 054683
14226-008EA 797.35 USD
14226-008 14226-012 60361-724 60361-728 60361-730 10799-632
HT8 GC Capillary Columns, SGE
Chromatography Columns
Innovative phase technologies and fused silica production capabilities provide end-to-end separation solutions.
  • 8% phenyl polycarborane-siloxane
  • Preferred column for polychlorinated biphenyl (PCB) compounds
  • Separates PCB’s on ortho ring substitution as well as boiling point
  • Low bleed
Application Areas: PCB congener alayses, nitro-substituted aromatics, polynuclear aromatic hydrocarbons, pesticides/herbicides.
Certifications: RoHS compliant per Directive 2011/65/EU