SGE Syringes, Gastight Syringe with Valve, Trajan Scientific and Medical
Supplier: Trajan Scientific and Medical
004279 005279 006279 007279 008110 008170 008510 008775 008975 008160 008560 008770 008970 009472 009670 009770
14225-278EA 214.22 USD
14225-278 14225-310 14225-336 14225-362 14225-382 14225-386 14225-400 14225-406 14225-414 60361-198 60361-202 60361-206 60361-210 60361-214 60361-218 60361-222
SGE Syringes, Gastight Syringe with Valve, Trajan Scientific and Medical
Syringes Syringes for Chromatography
These inert syringes are ideal for liquid and gas samples
Syringes are constructed from borosilicate glass with a fluoropolymer resin-tipped plunger, and assembled with an interference fit between plunger tip and barrel to effect gas sealing properties. Plungers are interchangeable on syringes of the same volume. Luer-lock fittings are not slotted.
Note: Syringes can be autoclaved but the plunger should be removed and heated and cooled slowly.
Suitable for manual and dispensing/diluting applications. Dispensed volume is accurate to ±1%.
Ordering information: Needles not included.