General Purpose Manual Syringes, MicroVolume, SGE
Supplier: Trajan Scientific and Medical
001495 002003 002035 002105 002839 003000 003050 004000 004050 005050 005055 006000 006050 007000 007050 002005 001400 001450 002030 002033 002100 002130 002133 002135 002180 002400 002450 001000 001050 002050 002080 001100 002000
14225-150EA 138.98 USD
14225-150 14225-176 14225-180 14225-192 14225-228 14225-240 14225-242 14225-268 14225-270 14225-296 14225-298 14225-330 14225-332 14225-352 14225-354 14233-538 60361-044 60361-046 60361-070 60361-072 60361-074 60361-080 60361-082 60361-084 60361-090 60361-100 60361-102 60361-422 60361-424 60361-426 60361-428 60361-038 60361-068
General Purpose Manual Syringes, MicroVolume, SGE
Syringes Syringes for Chromatography
General purpose plunger-in-barrel microvolume syringes
  • Plunger Protection incorporates a guide and button reinforcement to minimize the risk of bent plungers
  • SuperfleX Flexible Plungers use a super elastic alloy plunger that will not bend or kink
  • Guided Plungers have an extended barrel and reinforced plunger that provides additional protection
  • Accuracy and reproducibility ±1%
Scale Length of 54.1 mm for 5 µL and 10 µL syringes; 60 mm for 25 µL to 500 µL syringes.
Each plunger is individually fitted for each microsyringe for perfect sealing.