Argyle™ Vascular Tourniquet Kit, Covidien
Supplier: Cardinal Health
8888585000
60351-142CS 276.07 USD
60351-142
Argyle™ Vascular Tourniquet Kit, Covidien
Tourniquets
The Argyle™ vascular tourniquet kit is a sterile, single use, disposable kit containing all necessary components to snare and secure catheters, cannulas and vascular shunts in place in the surgical field.
  • Used to secure catheters, cannulas and vascular shunts in vessels
  • Retaining plug replaces hemostats which clutter field
  • Obturator is versatile, effective and easy to use, grasps umbilical tape and silicone loops
Sheath eliminates the need for cutting and sterilizing red rubber catheter segments. The sheath can be cut to further reduce clutter in the surgical field. The kit is complete, packaged sterile and ready to use, no assembly, preparation or sterilization required.
Delivery information: Includes red rubber sheath with retaining plug, an obturator and umbilical tape.