VWR® Talon® Support
Supplier: VWR International
60100-105
60100-105EA 513.3 USD
60100-105
VWR® Talon® Support
Support Stands
Multi-purpose support was originally designed to permit cylindrical vessels up to 305mm (12") in diameter to sit close to the upright rod.
  • Versatile
  • Heavy-duty design
  • Multi-purpose application
  • Supplied with stainless steel rod
  • Finished in black crackle lacquer
Support is ideal for mounting stirring motors or for heavy equipment. Each of the four corners rests on a rubber cushion. Supplied with stainless steel rod 457mm (18") high, 16mm (5/8") diameter.