VWR® Talon® End-to-End Connector
Supplier: VWR International
80862645
60079-100CS 860.77 USD
60079-100
VWR® Talon® End-to-End Connector
Support Rods and Frames
Strong aluminum alloy extends the length of lab frame rods.
  • Precisely bored openings
  • Strong aluminum alloy construction
  • Resistant to corrosion
Precision bore ensures alignment of rods. Comes complete with two set screws. Grips rods up to 13 mm (1/2").