Synthware Stopcock, High Vacuum, 1:10 Glass Plug, Kemtech America
Supplier: Kemtech America
S331230 S332044
60005-150EA 67.68 USD
60005-150 60005-152 60005-154
Synthware Stopcock, High Vacuum, 1:10 Glass Plug, Kemtech America
Stopcocks
High vacuum stopcock.
  • Straight bore
  • Heavy wall borosilicate construction
  • All glass stopcock
The 1:10 taper straight bore glass plug is precision ground and hand-lapped with corresponding numbers on the barrel and plug. Stopcocks are supplied with a stainless steel constant pressure retention spring and clip.