Synthware Stopcock, Teflon®, 120° Bore
Supplier: Kemtech America
S411530
60005-144EA 108.47 USD
60005-144
Synthware Stopcock, Teflon®, 120° Bore
Stopcocks
A (1:5 taper) Teflon® stopcock with three sidearms sealed 120° apart.
  • Heavy wall construction
  • 33 expansion borosilicate glass
Plug has a bore to allow connection of any two arms at once.