Spherical Pinch Clamps, Kemtech America
Supplier: Kemtech America
CC0012
60004-972EA 16.55 USD
60004-972
Spherical Pinch Clamps, Kemtech America
Clamps Standard Taper and Spherical Joint Clamps
Kemtech pinch clamp for spherical joints.
  • Stainless steel construction
  • Adjustable metal tensioner
For use with 12/5 ball joint, or #5 O-ring joints.