Glass Cutter, Natural Diamond, Kemtech America
Supplier: Kemtech America
DC18001
60004-970EA 74.12 USD
60004-970
Glass Cutter, Natural Diamond, Kemtech America
Cutters
This cutter is designed with rack on shank for breaking off single- or double-strength glass.
  • Extra-large natural diamond for durability
Handheld plate glass cutter has a durable diamond tip for extended life.