Synthware Springs
Supplier: Kemtech America
TH006 TH025 TH032
60004-940EA 2.2 USD
60004-940 60004-944 60000-758
Synthware Springs
Springs
Stainless steel retention springs.
  • Chemically resistant stainless steel
  • Compatible with other brands
Stainless steel springs for use with glassware that features hooks.