Synthware Check Valve
Supplier: Kemtech America
V180008
60004-922EA 67.62 USD
60004-922
Synthware Check Valve
Valves Check Valves
Gound seat for liquid seal.
  • Heavy wall borosilicate construction
  • All glass
  • Ground seat to ensure liquid seal
Heavy wall check valve is designed to prevent gas or liquid backflow.