Synthware Stopper with #15 O-Ring Joint, Kemtech America
Supplier: Kemtech America
S480015
60004-852EA 22.97 USD
60004-852
Synthware Stopper with #15 O-Ring Joint, Kemtech America
Stoppers Glass Stoppers
Designed for use with manifolds equipped with #15 O-rings.
  • Heavy wall borosilicate construction
  • Ground glass joints
  • #15 O-ring
Supplied complete with one #116 Viton* O-ring.