Synthware Storage Flask, #15 O-Ring Joint
Supplier: Kemtech America
S521000
60004-700EA 159.5 USD
60004-700
Synthware Storage Flask, #15 O-Ring Joint
Flasks Nitrogen Flasks
Storage Flask features a #15 O-ring joint on the sidearm and a high vacuum valve to control gas and/or liquid flow into the vessel.
  • Heavy wall construction
  • 33 expansion borosilicate glass
  • Standard ground glass joints work with other brands
Supplied complete with one #116 Viton® O-ring, and one high vacuum valve.