Synthware Connecting Inlet Adapter, Kemtech America
Supplier: Kemtech America
A560001
60004-662EA 115.64 USD
60004-662
Synthware Connecting Inlet Adapter, Kemtech America
Adapters Adapters, Joints with Tube Connectors
Adapter has an O-ring joint with a vacuum valve at a 90° angle connected to a piece of medium wall tubing.
  • O-ring joint
  • Vacuum valve at 90° angle
  • Medium wall tubing
  • One #116 Viton® O-ring included
Featuring a #15 O-ring joint with a 0-4mm high vacuum valve at a 90° angle connected to a piece of 10mm O.D. medium wall tubing. Overall height is 100mm. Each adapter is supplied complete with one #116 Viton® O-ring.