Synthware Vacuum Trap with #15 O-Ring Joint, Kemtech America
Supplier: Kemtech America
V325045
60004-630EA 199.88 USD
60004-630
Synthware Vacuum Trap with #15 O-Ring Joint, Kemtech America
Vacuum Traps
Vacuum trap is supplied with two #116 Viton® O-rings.
  • Heavy wall borosilicate construction
  • 35/20 spherical joint
Accessory items must be ordered separately. The inner and side tube O.D. is 18 mm. Note: Trap length is measured below the joint