Synthware Vacuum Trap with 35/20 Ball Joints, Kemtech America
Supplier: Kemtech America
V415045
60004-628EA 229.96 USD
60004-628
Synthware Vacuum Trap with 35/20 Ball Joints, Kemtech America
Vacuum Traps
Designed for use with vacuum manifolds having 35/20 socket joints.
  • Heavy wall borosilicate construction
  • 35/20 spherical joint
  • Adapters sold separately
Pump adapter and socket clamp must be ordered separately. Note: Trap length is measured below the joint.