Synthware Double Manifold with High Vacuum Valve and 35/20 Spherical Joint, Kemtech America
Supplier: Kemtech America
M360004
60004-590EA 920.93 USD
60004-590
Synthware Double Manifold with High Vacuum Valve and 35/20 Spherical Joint, Kemtech America
Manifolds
Double manifold is designed to incorporate a 0 - 8 mm high vacuum valve as the vacuum shut-off.
  • Hollow Glass stopcocks (Held in by Vacuum)
  • Heavy wall Borosilicate construction
  • 4 Ports
The front tube is intended for inert gases (17 mm O.D.), and features a 10 mm hose connection. The rear tube is intended for vacuum use (22 mm O.D.), and features two 35/20 socket joints. All components must be ordered separately.