Synthware Heat Transfer Block for Round Bottom Flasks, Microscale, Kemtech America
Supplier: Kemtech America
H200003
60004-230EA 90.89 USD
60004-230
Synthware Heat Transfer Block for Round Bottom Flasks, Microscale, Kemtech America
Block Heaters Blocks
Aluminum heat transfer block, used in place of a glass sand bath in microscale experiments.
  • Aluminum heat transfer block
  • Hold 3 flask sizes
Block holds 5 ml, 10 ml, and 25 ml round bottom flasks, with one hole for a standard thermometer.