Synthware Heat Transfer Block for Minivials, Microscale, Kemtech America
Supplier: Kemtech America
H200001
60004-228EA 87.91 USD
60004-228
Synthware Heat Transfer Block for Minivials, Microscale, Kemtech America
Block Heaters Blocks
Aluminum heat transfer block, used in place of a glass sand bath in microscale experiments.
  • Aluminum heat transfer block
  • Hold three sizes of vials
Block has two holes for 3 ml and 5 ml heavy wall reaction vials, four holes for Craig recrystallization tubes, and one hole for a standard thermometer.