Synthware Condenser, Air Reflux, Microscale, Kemtech America
Supplier: Kemtech America
C221420
60004-122EA 44.52 USD
60004-122
Synthware Condenser, Air Reflux, Microscale, Kemtech America
Condensers
Air reflux condenser for microscale applications.
  • Heavy wall borosilicate construction
  • Ground glass joint
Length between ST joints: 96 mm.