Synthware Adapter, Claisen, Threaded, Microscale, Kemtech America
Supplier: Kemtech America
A371410
60004-112EA 54.93 USD
60004-112
Synthware Adapter, Claisen, Threaded, Microscale, Kemtech America
Distillation Components Distillation Adapters
Claisen joint adapter for microscale applications
  • Threaded
  • For microscale applications
  • Two compression caps and two O-rings included
Threaded Claisen joint adapter is supplied with two nylon compression caps and two Viton® O-rings.
Supplied with two nylon compression caps and two Viton® O-rings.