Synthware Adapter, Inlet or Thermometer, Microscale, Kemtech America
Supplier: Kemtech America
A771410
60004-104EA 35.53 USD
60004-104
Synthware Adapter, Inlet or Thermometer, Microscale, Kemtech America
Adapters Adapters, Joints with Tube Connectors
This is a joint adapter suitable for microscale applications.
  • Vacuum-tight seal
  • Cap accommodates thermometer
  • Nylon compression cap and Viton® O-ring included
Supplied with a nylon compression cap and Viton® O-ring to form a vacuum tight seal. To use as a syringe port, replace the O-ring with PTFE -lined septum. Cap accommodates a 5.0 to 7.5 mm ext. Ø thermometer.