Synthware Adapter, Vacuum, with Stopcock, Microscale, Kemtech America
Supplier: Kemtech America
A381410
60004-102EA 81.42 USD
60004-102
Synthware Adapter, Vacuum, with Stopcock, Microscale, Kemtech America
Adapters Adapters, Conical Ground Joints
Vacuum adapters are used to regulate the flow of gases or liquids into reation systems.
  • With stopcock
  • For microscale applications
Vacuum joint adapter features a 1:5 PTFE stopcock.