Synthware Adapter, Gas Inlet/Outlet, Microscale, Kemtech America
Supplier: Kemtech America
A301410
60004-100EA 23.24 USD
60004-100
Synthware Adapter, Gas Inlet/Outlet, Microscale, Kemtech America
Adapters Adapters, Joints with Tube Connectors
Gas inlet/outlet joint adapter is suitable for microscale applications.
  • For gas inlet/outlet
  • [ST] joint
Adapter is equipped with a standard taper joint.